missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMSN1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-976
This item is not returnable.
View return policy
Description
SAMSN1 Polyclonal specifically detects SAMSN1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| SAMSN1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| HACS1gene with homology to KIAA0790 protein10NASH1, Hematopoietic adaptor containing SH3 and SAM domains 1, SAM and SH3 domain containing 1, SAM and SH3 domain containing 2, SAM domain, SH3 domain and nuclear localisation signals, 1, SAM domain, SH3 domain and nuclear localization signals 1, SAM domain, SH3 domain and nuclear localization signals protein 1, SAM domain-containing protein SAMSN-1, SH3D6B, SH3-SAM adaptor protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Please note that the immunogen sequence is present in NP_071419.2 (and in AAK07746) but is no longer present in the latest reference sequence (NP_071419.3 at 05/10/05). This latest version has not been subject to final review by NCBI. This antibody may be recognising one particular isoform. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| NP_071419.2 | |
| SAMSN1 | |
| Peptide with sequence IRLDTLSASLGRSC corresponding to N-Terminus according to NP_071419.2. | |
| 0.1 mg | |
| Stem Cell Markers | |
| 64092 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction