missing translation for 'onlineSavingsMsg'
Learn More

SAMSN1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18419459 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419459 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419459 Supplier Novus Biologicals Supplier No. NBP297172JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SAMSN1 Polyclonal antibody specifically detects SAMSN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAMSN1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias HACS1gene with homology to KIAA0790 protein10NASH1, Hematopoietic adaptor containing SH3 and SAM domains 1, SAM and SH3 domain containing 1, SAM and SH3 domain containing 2, SAM domain, SH3 domain and nuclear localisation signals, 1, SAM domain, SH3 domain and nuclear localization signals 1, SAM domain, SH3 domain and nuclear localization signals protein 1, SAM domain-containing protein SAMSN-1, SH3D6B, SH3-SAM adaptor protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SAMSN1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 64092
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.