missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress SAMS

Product Code. 16481828
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481828 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16481828 Supplier MedChemExpress Supplier No. HYP0136

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAMS peptide is a specific substrate for the AMP-activated protein kinase (AMPK).

Specifications

Purity 99.88%
CAS 125911-68-4
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Metabolism-sugar/lipid metabolism
Molecular Weight (g/mol) 1779.15 g/mol
Product Type Peptides
Quantity 5 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (56.21 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.