Learn More
Thermo Scientific™ Sampling Manifold Accessory for HyPerforma™ Single-Use Bioreactors, with Luer locks
GREENER_CHOICE
Product Code. 31940829 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
50/100 mL
Quantity:
1 Ea.
10/Pk.
Unit Size:
Pack of 10
Pack of 5
Pack of 60
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Quantity Unit Size
31940829 50/100 mL 10/Pk. Pack of 60
17889756 50/100 mL 1 Ea. Pack of 10
30040085 50/100 mL 1 Ea. Pack of 5
17809766 50/100 mL 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31940829 Supplier Thermo Scientific™ Supplier No. SH31263.02
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Sterile Sampling Manifold is an accessory for the HyPerforma Single-Use Bioreactors and is available to weld onto the sample line to take sample sets.
The Sterile Sampling Manifold is an accessory for the HyPerforma Single-Use Bioreactors and is available to weld onto the sample line to take sample sets.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50/100 mL
Quantity 10/Pk.
Tubing Style C-Flex