Learn More
Thermo Scientific™ Sampling Manifold Accessory for HyPerforma™ Single-Use Bioreactors, with Luer locks
Product Code. 17809766 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
50/100 mL
Quantity:
1 Ea.
10/Pk.
Unit Size:
Pack of 10
Product Code. 17809766
Brand: Thermo Scientific™ undefined
The Sterile Sampling Manifold is an accessory for the HyPerforma Single-Use Bioreactors and is available to weld onto the sample line to take sample sets.
The Sterile Sampling Manifold is an accessory for the HyPerforma Single-Use Bioreactors and is available to weld onto the sample line to take sample sets.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50/100 mL
Quantity 10/Pk.
Tubing Style C-Flex