missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Sample Splitters

Product Code. 10379870 Shop All Retsch Products
Change view
Click to view available options
Depth (Metric):
6.3 mm
12.5 mm
25 mm
37.5 mm
50 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) unitSize
10379870 6.3 mm Each
10572121 50 mm Each
10562121 25 mm Each
10432691 12.5 mm Each
10339780 37.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10379870 Supplier RETSCH Supplier No. 40.610.0001
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Product Type Sample Splitter
Depth (Metric) 6.3 mm
Includes 3 Collecting Receptacles 1.5L Each of Tin Plate, Stand of Coated Sheet Steel and Dividing Head Made of Stainless Steel
Particle Size 6.3 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.