Learn More
Biochrom Sample Plate Replacement Kit
Product Code. 11330322
Change view
Click to view available options
For Use With (Equipment):
NanoVue Plus Spectrophotometer
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
11330322 NanoVue Plus Spectrophotometer Each
1 options
Product Code. 11330322 Supplier Biochrom Supplier No. 28955301
Only null left
Add to basket
Add to basket
Sample Plate Replacement Kit for use with NanoVue Plus Spectrophotometer.
  • The sample plates can be easily and quickly replaced should this ever become necessary
  • Sample plates can be clipped in and out by the user
  • Pathlength calibration must be performed following a change of plate
Specifications
Display No Display
For Use With (Equipment) NanoVue Plus Spectrophotometer
Shape Rectangular