Learn More
VICI Valco Instruments™ PEEK Loops for Rheodyne™ Valves
Product Code. 12720924
Product Code. 12720924
Brand: VICI Valco Instruments™ 724JRSLR2204
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PEEK sample loops are particularly well suited for separations involving proteins, peptides, nucleic acids or other samples of biological origin as well as in IC systems where metal traces may interact with ionic samples or release transition metals that may deactivate ion exchange columns.
  • Fits all Rheodyne metal and metal-free valves
  • Wide range of volumes available
  • Biocompatible
  • Made of premium grade PEEK tubing
  • PEEK is a mechanically strong, chemically inert polymer ideal for HPLC applications.
  • Since our PEEK sample loops contain no metals or other extractable compounds, no foreign materials will be introduced into the system.
  • Softener-free polyethylene caps are used to avoid contamination of the tubing.
Specifications
Max. Pressure 4500 psi
Volume (Metric) 100 μL