missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Sample Cups for Roche Cobas Mira

Product Code. 31922588
Change view
Click to view available options
Color:
Green
Unit Size:
Pack of 12000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31922588 Green Pack of 12000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31922588 Supplier Globe Scientific Supplier No. 6500G
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use on automated instrumentation including the Cobas Mira, Bio, Fara and Horiba ABX analyzers. Globe Scientific Sample Cups for Roche Cobas Mira are constructed of high density polyethylene (HDPE) and feature an attached snap cap.

  • For use on automated instrumentation including the Cobas Mira, Bio, Fara and Horiba ABX analyzers
  • Attached snap cap
  • High Density Polyethylene (HDPE)
  • Green (not pictured)

Specifications

For Use With (Equipment) Cobas Mira, Bio, Fara and Horiba ABX Analyzers
Color Green
Material HDPE
Product Type Sample Cup
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.