missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Sample Cups for Abbot Architect Series Analyzers

Product Code. 31923167 Shop All Globe Scientific Products
Change view
Click to view available options
Quantity:
1000
Unit Size:
Pack of 5000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31923167 1000 Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31923167 Supplier Globe Scientific Supplier No. 5106
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use with the Abbott Architect Series Analyzers. Globe Scientific Sample Cups for Abbot Architect Series Analyzers are constructed from polystyrene and are specifically designed for use with these products.

  • For use with the Abbott Architect Series Analyzers
  • Specifically designed for use with these products
  • Polystyrene (PS) construction, natural color

Specifications

Type Sample Cup
Color Natural
Quantity 1000
Material Polystyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.