Learn More
Fisherbrand™ Polyethylene Sample Bag
For use in the laboratory to take samples into a sterile environment.
Brand:  Fisherbrand™ 14955193
Product Code. 11768026
  • £72.85 / Pack of 250
Only null left
Estimated Shipment: 20-05-2026
Add to basket
Description
Description
  • Secure, contaminant free bags to ensure dependable analysis results
  • Made from heavy gauge, virgin polyethylene that meets FDA specifications
  • Can accommodate solid, semi-solid and liquid materials
  • Easy to use, insert the sample and seal by twirling the bag outward four times and bending wire ends inward
  • Flat wire closures provide extra strength to seal larger and heavier samples