missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMHD1 Rabbit anti-Human, Mouse, Rat, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | SAMHD1 |
|---|---|
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | CoraLite 594 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17761379
|
Proteintech
CL594-12586-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17679926
|
Proteintech
CL594-12586-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SAMHD1 Polyclonal antibody apecifically detetects SAMHD1 antigen in Human, Mouse, Rat Samples. It is validated for Flow Cytometry.Specifications
| SAMHD1 | |
| Polyclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| BC036450 | |
| SAMHD1 | |
| IgG | |
| Antigen affinity purification |
| Flow Cytometry | |
| CoraLite 594 | |
| Rabbit | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 25939 | |
| SAMHD1 fusion protein Ag3287 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title