missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAMHD1 Monoclonal Antibody (OTI2D7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525299
This item is not returnable.
View return policy
Description
SAMHD1 Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene may play a role in regulation of the innate immune response. The encoded protein is upregulated in response to viral infection and may be involved in mediation of tumor necrosis factor-alpha proinflammatory responses. Mutations in this gene have been associated with Aicardi-Goutieres syndrome.
Specifications
| SAMHD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q60710, Q9Y3Z3 | |
| SAMHD1 | |
| Full length human recombinant protein of SAMHD1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| OTI2D7 | |
| Unconjugated | |
| SAMHD1 | |
| CHBL2; DCIP; Dendritic cell-derived IFNG-induced protein; deoxynucleoside triphosphate triphosphohydrolase SAMHD1; dNTPase; E330031J07Rik; EGK_02438; HDDC1; hSAMHD1; IFN-gamma induced; interferon-gamma-inducible protein Mg11; Mg11; Mg21; Monocyte protein 5; MOP5; MOP-5; mSAMHD1; RP1-132F21.1; SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1; SAM domain and HD domain 1; SAM domain and HD domain, 1; SAM domain and HD domain-containing protein 1; Samhd1; SBBI88 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 25939, 56045 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction