missing translation for 'onlineSavingsMsg'
Learn More

SAMD9 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA554378

Additional Details : Weight : 0.02000kg

Product Code. 15742311

  • £427.00 / 100µL
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: VSQKERRETS KQKQKGKENP DMANPSAMST TAKGSKSLKV ELIEDKIDYT KERQPSIDLT CVSYPFDEFS NPYRYKLDFS LQP Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 49%.

Observed MW of SAMD9 is from 148 kDa.
Specifications

Specifications

SAMD9
Polyclonal
Unconjugated
SAMD9
C7orf5; DRIF1; expressed in aggressive fibromatosis; KIAA2004; NFTC; OEF1; OEF2; RGD1563091; SAM domain-containing protein 9; SAMD9; similar to OEF2; sterile alpha motif domain containing 9; sterile alpha motif domain-containing protein 9
Rabbit
Antigen affinity chromatography
RUO
54809
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5K651
SAMD9
Recombinant protein corresponding to Human SAMD9
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only