missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SAMD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £532.00
Specifications
| Antigen | SAMD8 |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066992
|
Novus Biologicals™
NBP3-30871-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066834
|
Novus Biologicals™
NBP3-30871-100ug |
100 μg |
£532.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SAMD8 Polyclonal antibody specifically detects SAMD8 in Human samples. It is validated for ELISA,Western BlotSpecifications
| SAMD8 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| Phosphate buffered saline | |
| 142891 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 5-10 μg/mL, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ25082, SAM domain-containing protein 8, SMSr, sphingomyelin synthase related, sphingomyelin synthase-related protein 1, sterile alpha motif domain containing 8, Sterile alpha motif domain-containing protein 8 | |
| Full-length recombinant SAMD8 protein | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title