missing translation for 'onlineSavingsMsg'
Learn More

SAMD4B Antibody (PCRP-SAMD4B-1H3), FITC, Novus Biologicals™

Product Code. 18344955 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344955 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344955 Supplier Novus Biologicals Supplier No. NBP314152F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SAMD4B Monoclonal antibody specifically detects SAMD4B in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAMD4B
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-SAMD4B-1H3
Conjugate FITC
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias FLJ10211, hSmaug2, MGC99832, protein Smaug homolog 2, SAM domain-containing protein 4B, smaug 2, smaug homolog B, SMAUG2, SMGB, sterile alpha motif domain containing 4B, sterile alpha motif domain-containing protein 4B
Host Species Mouse
Immunogen Recombinant full-length humanSAMD4Bprotein (Uniprot: Q5PRF9)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55095
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.