missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SAMD4A 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Target | SAMD4A |
| Synonym | DKFZp434H0350, DKFZp686A1532, hSmaug1DKFZP434H0350, KIAA1053SMAUG1, protein Smaug homolog 1, SAM domain-containing protein 4A, SAMD4, Smaug, Smaug 1, smaug homolog, Smaug1, SMG, SMGA, sterile alpha motif domain containing 4, sterile alpha motif domain containing 4A, Sterile alpha motif domain-containing protein 4A |
| Gene Symbol | SAMD4A |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction