missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Samco™ Transfer Pipettes
GREENER_CHOICE

Product Code. 15377883
Change view
Click to view available options
Volume (Metric):
0.6 mL
1.05 mL
1.8 mL
3.1 mL
4.9 mL
5.5 mL
7.5 mL
7.7 mL
8.0 mL
8.6 mL
9.5 mL
Sterility:
Non-sterile
Nonsterile
Sterile
Bulb Draw (Metric):
0.3 mL
0.9 mL
1.05 mL
1.8 mL
2.2 mL
2.4 mL
2.8 mL
3.2 mL
3.5 mL
4.6 mL
6.0 mL
6.2 mL
Packaging Type:
20/Bag, 25 Bags/Pk, 8 Pks/Cs.
20/Bag, 400/Pk
20/Bag, 500/Pk
4000/Cs
Bulk 250/Pk
Bulk 400/Pk
Bulk 500/Pack
Bulk 500/Pk
Bulk, 400/Pk.
Bulk, 500/pk
Individually Wrapped 250/Pk
Individually Wrapped 400/Pk
Individually Wrapped 500/Pk
Individually Wrapped, 400/Pk
Individually Wrapped, 500/pk
Unit Size:
Pack of 250
Pack of 400
Pack of 4000
Pack of 500
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Bulb Draw (Metric) Packaging Type unitSize
15377883 4.9 mL Nonsterile 1.8 mL Bulk 500/Pack Pack of 500
15307813 7.7 mL Non-sterile 3.5 mL Bulk 500/Pk Pack of 500
15337813 8.6 mL Sterile 6.0 mL Individually Wrapped, 400/Pk Pack of 400
15307913 5.5 mL Non-sterile 2.4 mL Bulk 500/Pk Pack of 500
11661839 7.7 mL Non-sterile 3.2 mL Bulk, 500/pk Pack of 500
15778257 7.7 mL Sterile 3.2 mL 4000/Cs Pack of 4000
11367873 7.7 mL Sterile 3.2 mL Individually Wrapped, 500/pk Pack of 500
12394508 8.0 mL Non-sterile 4.6 mL Bulk 400/Pk Pack of 400
15327883 3.1 mL Non-sterile 2.2 mL Bulk 400/Pk Pack of 400
15317813 7.7 mL Sterile 3.5 mL Individually Wrapped 500/Pk Pack of 500
15327813 7.7 mL Sterile 3.5 mL 20/Bag, 25 Bags/Pk, 8 Pks/Cs. Pack of 500
10354470 8.6 mL Non-sterile 6.0 mL Bulk 400/Pk Pack of 400
15357813 8.6 mL Sterile 6.0 mL 20/Bag, 400/Pk Pack of 400
15252089 1.8 mL Non-sterile 0.9 mL Bulk 500/Pk Pack of 4000
15337883 7.5 mL Non-sterile 2.8 mL Bulk 400/Pk Pack of 400
15317863 1.8 mL Sterile 0.9 mL Individually Wrapped 500/Pk Pack of 500
15327863 1.8 mL Sterile 0.9 mL 20/Bag, 500/Pk Pack of 500
15357883 7.5 mL Sterile 2.8 mL Individually Wrapped 400/Pk Pack of 400
15367883 7.5 mL Sterile 2.8 mL 20/Bag, 500/Pk Pack of 500
15397883 4.9 mL Sterile 1.8 mL Individually Wrapped 500/Pk Pack of 500
15307893 4.9 mL Sterile 1.8 mL 20/Bag, 500/Pk Pack of 500
15337893 9.5 mL Non-sterile 6.2 mL Bulk 250/Pk Pack of 250
15347893 9.5 mL Sterile 6.2 mL Individually Wrapped 250/Pk Pack of 250
15357893 9.5 mL Sterile 6.2 mL 20/Bag, 400/Pk Pack of 400
15307883 1.05 mL Non-sterile 1.05 mL Bulk 500/Pk Pack of 500
15307873 0.6 mL Non-sterile 0.3 mL Bulk, 500/pk Pack of 500
15317883 3.1 mL Non-sterile 2.2 mL Bulk, 400/Pk. Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
Product Code. 15377883 Supplier Thermo Scientific™ Supplier No. 336
Add to basket
Edge
Add to basket

Prevent cross-contamination with these transfer pipettes, which have a built-in bulb that eliminates the need for plugging.

Prevent cross-contamination with Thermo Scientific™ Samco™ Transfer Pipettes, which have a built-in bulb that eliminates the need for plugging, packaging and autoclaving. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.

Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel. Molded in one piece from see-through low-density polyethylene.

  • Capacity: approximately 5 mL
  • Delivers 24 to 26 drops/mL
  • Unbreakable all-in-one pipettes eliminate the hazards of broken glass and exposure to infectious materials
  • Inert to biological fluids and most acids
  • Work well anywhere there is a need for quick, safe transfer of fluids
  • One squeeze draws from 2.0 to 3.3 mL (depending on size used)
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)
TRUSTED_SUSTAINABILITY

Specifications

Bulb Draw (Metric) 1.8 mL
Material LDPE
Sterility Nonsterile
Drops/mL 23
For Use With (Application) Research
Graduated No
Quantity 500/Pk.
Packaging Type Bulk 500/Pack
Volume (Metric) 4.9 mL
Length (Metric) 15.4 cm
Type General Purpose Transfer Pipet
Disposable Single-use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.