missing translation for 'onlineSavingsMsg'
Learn More

Samarium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)

Product Code. 11323848
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
Unit Size:
100mm
10mm
25mm
50mm
This item is not returnable. View return policy

Product Code. 11323848

Brand: Thermo Scientific Alfa Aesar 040297.DH

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-19-9 | Sm | 150.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Specifications

Physical Form Cast Rod
Quantity 25 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Assay (metals basis excluding Ta)
Chemical Name or Material Samarium rod, 6.35mm (0.25 in.) dia.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.