missing translation for 'onlineSavingsMsg'
Learn More

Samarium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)

Product Code. 11363848
Change view
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
Unit Size:
100mm
10mm
25mm
50mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363848 100 mm 100mm
11383848 10 mm 10mm
11353848 25 mm 25mm
11373848 50 mm 50mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11363848 Supplier Thermo Scientific Alfa Aesar Supplier No. 040298.DM
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-19-9 | Sm | 150.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Specifications

Quantity 100 mm
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.96g/mm
Form Cast Rod
Percent Purity 99.9%
Assay (metals basis excluding Ta)
Chemical Name or Material Samarium rod, 12.7mm (0.5 in.) dia.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.