missing translation for 'onlineSavingsMsg'
Learn More

Samarium pieces, sublimed dendritic, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11371789
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7440-19-9
Sm
150.36
MFCD00011233 MFCD00151299
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
23951
CHEBI:33374
samarium
[Sm]
This item is not returnable. View return policy
7440-19-9
Sm
150.36
MFCD00011233 MFCD00151299
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
23951
CHEBI:33374
samarium
[Sm]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color Gray
UN Number UN3178
Quantity 10 g
Percent Purity 99.9%
Odor Odorless
Form Pieces
Assay (REO)
Chemical Name or Material Samarium pieces, Sublimed dendritic

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'