missing translation for 'onlineSavingsMsg'
Få mere at vide

Samarium pieces, 12mm (0.47in) & down, 99.9% (REO)

Artikelnummer. 11386587
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
1 g
10 g
50 g
Pakningsstørrelse:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11386587 10 g 10g
11396587 1 g 1g
11306597 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11386587 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. 000609.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

CAS: 7440-19-9 | Sm | 150.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in air stable poly bottle
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Tekniske data

Color Gray
Quantity 10 g
Odor Odorless
Packaging Poly bottle
Particle Size 12mm (0.47 in.) & down
Sensitivity Air and moisture sensitive
Form Pieces
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Samarium pieces

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.