missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) trifluoromethanesulfonate, 98%

Product Code. 11399106
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399106 5 g 5g
11309116 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399106 Supplier Thermo Scientific Alfa Aesar Supplier No. L20253.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52093-28-4 | C3F9O9S3Sm | 597.55 g/mol

Used as catalyst for Baylis-Hillman reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as catalyst for Baylis-Hillman reaction.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in a cool, dry place in a tightly closed containers. Stable under recommended storage conditions. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52093-28-4
Molecular Formula C3F9O9S3Sm
Molecular Weight (g/mol) 597.55
MDL Number MFCD00192431
InChI Key DDCWGUIPLGMBPO-UHFFFAOYSA-K
Synonym samarium iii trifluoromethanesulfonate, samarium triflate, samarium iii triflate, samarium trifluoromethanesulfonate, trifluoromethanesulfonic acid samarium iii salt, samarium iii trifluoromethanesulfonate 97+%, sm otf 3, 3cf3o3s.sm, samarium 3+ tritriflate
PubChem CID 10145316
IUPAC Name samarium(3+);trifluoromethanesulfonate
SMILES [Sm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 597.56
Percent Purity 98%
Chemical Name or Material Samarium(III) trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.