missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) oxide, REacton™, 99.99% (REO)

Product Code. 11386278
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11386278

Brand: Thermo Scientific Alfa Aesar 011230.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12060-58-1 | Sm2O3

Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. It finds use as a neutron absorber in nuclear power reactors to control the reaction. It is also used as a catalyst for the dehydration of primary alcohols to aldehydes or ketones. It forms samarium perchlorate when treated with perchloric acid, which is used as a reference material for checking the wavelength scale of a spectrophotometer in the range of 200 to 500 nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-58-1
Molecular Formula Sm2O3
MDL Number MFCD00011237

Specifications

Melting Point 2300°C
Color Violet
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water.
Formula Weight 348.7
Refractive Index 1.97
Appearance Off-white powder
Density 8.347g/cm3 at 20°C
Chemical Name or Material Samarium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.