missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) oxalate hydrate, REacton™, 99.97% (REO), Thermo Scientific™

Product Code. 11372268
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11372268

Brand: Thermo Scientific Alfa Aesar 013176.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14175-03-2
Molecular Formula C6H26O22Sm2
Molecular Weight (g/mol) 750.97
MDL Number MFCD00150489
InChI Key IKPDQYODFRQYAD-UHFFFAOYSA-N
Synonym samarium iii oxalate hydrate 2:3:10, ethanedioic acid,samarium 3+ salt, hydrate 3:2:10
PubChem CID 121488066
SMILES O.O.O.O.O.O.O.O.O.O.[Sm].[Sm].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O

Specifications

Physical Form Powder
Quantity 100 g
Linear Formula Sm2(C2O4)3·xH2O
UN Number UN3288
IUPAC Name oxalic acid;samarium;decahydrate
Formula Weight 564.78 (Anhydrous)
Percent Purity 99.97%
Grade Reacton
Assay (REO)
Chemical Name or Material Samarium(III) oxalate hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.