missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) oxalate hydrate, REacton™, 99.97% (REO), Thermo Scientific™

Product Code. 11382268
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382268 25 g 25g
11392268 5 g 5g
11372268 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11382268 Supplier Thermo Scientific Alfa Aesar Supplier No. 013176.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14175-03-2
Molecular Formula C6H26O22Sm2
Molecular Weight (g/mol) 750.97
MDL Number MFCD00150489
InChI Key IKPDQYODFRQYAD-UHFFFAOYSA-N
Synonym samarium iii oxalate hydrate 2:3:10, ethanedioic acid,samarium 3+ salt, hydrate 3:2:10
PubChem CID 121488066
SMILES O.O.O.O.O.O.O.O.O.O.[Sm].[Sm].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O

Specifications

Physical Form Powder
Quantity 25 g
Linear Formula Sm2(C2O4)3·xH2O
UN Number UN3288
IUPAC Name tris(oxalic acid) decahydrate disamarium
Formula Weight 564.78 (Anhydrous)
Percent Purity 99.97%
Grade Reacton
Assay (REO)
Chemical Name or Material Samarium(III) oxalate hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.