missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

£40.40 - £335.00

Chemical Identifiers

CAS 13759-83-6
Molecular Formula H12N3O15Sm
Molecular Weight (g/mol) 444.46
MDL Number MFCD00149857
InChI Key HDCOFJGRHQAIPE-UHFFFAOYSA-N
Synonym samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
PubChem CID 203081
SMILES O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11376268
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011224.06
5 g
£40.40
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11396268
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011224.14
25 g
£121.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11386268
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011224.22
100 g
£335.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Samarium(III) nitrate hydrate is used as a catalyst in von Pechmann condensation for the preparation of coumarins. Further, it finds use in optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. It is also used as a laboratory reagent. In addition to this, it is used in NMR spectroscopy for spectral resolution and is used for doping purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13759-83-6
444.46
HDCOFJGRHQAIPE-UHFFFAOYSA-N
203081
H12N3O15Sm
MFCD00149857
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

78°C to 79°C
Yellow
Crystalline Aggregates
MFCD00149857
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
HDCOFJGRHQAIPE-UHFFFAOYSA-N
samarium(3+);trinitrate;hexahydrate
203081
99.99%
Odorless
2.375 g/mL
Samarium(III) nitrate hydrate
13759-83-6
∼3
H12N3O15Sm
UN1477
Soluble in water and acetonitrile.
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
444.46
336.36 (Anhydrous)
Reacton™
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P380+P375-P372-P380-P501c

H272-H315-H320-H335

missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II

missing translation for 'einecsNumber' : 233-798-6

missing translation for 'rtecsNumber' : VP3150000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only