missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) nitrate hydrate, REacton™, 99.99% (REO)

Product Code. 11386268
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386268 100 g 100g
11376268 5 g 5g
11396268 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11386268 Supplier Thermo Scientific Alfa Aesar Supplier No. 011224.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13759-83-6 | H12N3O15Sm | 444.46 g/mol

Samarium(III) nitrate hydrate is used as a catalyst in von Pechmann condensation for the preparation of coumarins. Further, it finds use in optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. It is also used as a laboratory reagent. In addition to this, it is used in NMR spectroscopy for spectral resolution and is used for doping purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13759-83-6
Molecular Formula H12N3O15Sm
Molecular Weight (g/mol) 444.46
MDL Number MFCD00149857
InChI Key HDCOFJGRHQAIPE-UHFFFAOYSA-N
Synonym samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
PubChem CID 203081
SMILES O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 78°C to 79°C
Color Yellow
pH ∼3
Physical Form Crystalline Aggregates
Quantity 100 g
UN Number UN1477
Solubility Information Soluble in water and acetonitrile.
IUPAC Name samarium(3+) hexahydrate trinitrate
Formula Weight 336.36 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Density 2.375 g/mL
Assay (REO)
Chemical Name or Material Samarium(III) nitrate hydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Samarium(III) nitrate hydrate, REacton
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Samarium(III) nitrate, hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.