missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) hydroxide hydrate, 99% (REO)

Product Code. 11328318
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328318 25 g 25g
11338318 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328318 Supplier Thermo Scientific Alfa Aesar Supplier No. 039512.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20403-06-9 | H3O3Sm | 201.38 g/mol

Samarium hydroxide (Sm(OH)3) is an important samarium compound with many potential applications in electronics, optics and catalysis. It is used as a chemical intermediate. It is used in the separation and preconcentration procedures for the determination of lead using spectrometric techniques. It has specialized uses in glass, phosphors, lasers, and thermoelectric devices and it is mainly applied in Neutron absorption.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20403-06-9
Molecular Formula H3O3Sm
Molecular Weight (g/mol) 201.38
MDL Number MFCD00050108
InChI Key BCYBEIXXOVNETJ-UHFFFAOYSA-K
IUPAC Name samarium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Sm+3]

Specifications

Physical Form Lump/Powder
Quantity 25 g
UN Number UN3262
Solubility Information Soluble in water and moderately soluble in strong mineral acids.
Formula Weight 201.37 (Anhydrous)
Percent Purity 99%
Assay (REO)
Chemical Name or Material Samarium(III) hydroxide hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.