Learn More
Samarium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals
£133.00 - £399.00
Chemical Identifiers
CAS | 13765-24-7 |
---|---|
Molecular Formula | F3Sm |
Molecular Weight (g/mol) | 207.36 |
MDL Number | MFCD00049540 |
InChI Key | OJIKOZJGHCVMDC-UHFFFAOYSA-K |
IUPAC Name | samarium(3+) trifluoride |
SMILES | [F-].[F-].[F-].[Sm+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346918
|
Thermo Scientific Alfa Aesar
013660.14 |
25 g |
£133.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356918
|
Thermo Scientific Alfa Aesar
013660.22 |
100 g |
£399.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Samarium(III) fluoride has specialized uses in glass, phosphors, lasers, and thermoelectric devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13765-24-7 | |
207.36 | |
OJIKOZJGHCVMDC-UHFFFAOYSA-K | |
[F-].[F-].[F-].[Sm+3] |
F3Sm | |
MFCD00049540 | |
samarium(3+) trifluoride |
Specifications
1306°C | |
Powder | |
MFCD00049540 | |
Soluble moderately in strong mineral acids. Insoluble in water. | |
[F-].[F-].[F-].[Sm+3] | |
207.36 | |
99.9% | |
Odorless | |
(REO) |
13765-24-7 | |
F3Sm | |
UN3288 | |
OJIKOZJGHCVMDC-UHFFFAOYSA-K | |
samarium(3+) trifluoride | |
207.35 | |
Reacton™ | |
Hygroscopic | |
Samarium(III) fluoride, Anhydrous |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 237-367-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only