missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Samarium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Samarium Acetate has specialized uses in glass, phosphors, lasers, and thermoelectric devices. Another important application of Samarium and its compounds is as catalyst and chemical reagent. One of the most important applications of Samarium is in Samarium-Cobalt magnets. . These magnets are found in small motors, headphones, and high-end magnetic pickups for guitars and related musical instruments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 300°C |
Physical Form | Crystalline Aggregates |
Quantity | 25 g |
Solubility Information | Soluble in water |
Formula Weight | 327.48 (Anhydrous) |
Percent Purity | 99.9% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 1.94 g/mL |
Assay | (REO) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'