missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) acetate hydrate, 99.9% (REO)

Product Code. 11382558
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382558 100 g 100g
11392558 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382558 Supplier Thermo Scientific Alfa Aesar Supplier No. 014604.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17829-86-6 | C6H9O6Sm | 327.49 g/mol

Samarium Acetate has specialized uses in glass, phosphors, lasers, and thermoelectric devices. Another important application of Samarium and its compounds is as catalyst and chemical reagent. One of the most important applications of Samarium is in Samarium-Cobalt magnets. . These magnets are found in small motors, headphones, and high-end magnetic pickups for guitars and related musical instruments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17829-86-6
Molecular Formula C6H9O6Sm
Molecular Weight (g/mol) 327.49
MDL Number MFCD00150124
InChI Key JPDBEEUPLFWHAJ-UHFFFAOYSA-K
IUPAC Name samarium(3+) triacetate
SMILES [Sm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Boiling Point 300°C
Physical Form Crystalline Aggregates
Quantity 100 g
Solubility Information Soluble in water
Formula Weight 327.48 (Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 1.94 g/mL
Assay (REO)
Chemical Name or Material Samarium(III) acetate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.