missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

Product Code. p-7051174
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11398588

Brand: Thermo Scientific Alfa Aesar 040665.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64438-53-5 | C15H21O6Sm | 447.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64438-53-5
Molecular Formula C15H21O6Sm
Molecular Weight (g/mol) 447.69
MDL Number MFCD00792904
InChI Key JTZJMDRBZZVNSP-UHFFFAOYSA-N
Synonym samarium 2,4-pentanedionate, samarium iii acetylacetonate hydrate, samarium iii 2,4-pentanedionate hydrate, samarium iii acetylacetonate hydrate trace metals basis, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy samario oxy pent-3-en-2-one hydrate
PubChem CID 16213768
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;samarium;hydrate
SMILES [Sm+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 5 g
Formula Weight 447.68 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Samarium(III) 2,4-pentanedionate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.