missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

Product Code. 11388588
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388588 25 g 25g
11398588 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388588 Supplier Thermo Scientific Alfa Aesar Supplier No. 040665.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64438-53-5 | C15H21O6Sm | 447.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64438-53-5
Molecular Formula C15H21O6Sm
Molecular Weight (g/mol) 447.69
MDL Number MFCD00792904
InChI Key JTZJMDRBZZVNSP-UHFFFAOYSA-N
Synonym samarium 2,4-pentanedionate, samarium iii acetylacetonate hydrate, samarium iii 2,4-pentanedionate hydrate, samarium iii acetylacetonate hydrate trace metals basis, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy samario oxy pent-3-en-2-one hydrate
PubChem CID 16213768
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;samarium;hydrate
SMILES [Sm+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 25 g
Formula Weight 447.68 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Samarium(III) 2,4-pentanedionate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.