missing translation for 'onlineSavingsMsg'
Learn More

Samarium(II) iodide, 0.07 - 0.12M in THF, stab., Thermo Scientific Chemicals

£73.80

Chemical Identifiers

CAS 32248-43-4
MDL Number MFCD00058873
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15471087
View Documents Promotion Details
Thermo Scientific Alfa Aesar
022360.AE
100 mL
£73.80
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32248-43-4
MFCD00058873

Specifications

32248-43-4
Ether-like
UN3271
404.21
Liquid
0.922 g/mL
MFCD00058873
Air and moisture sensitive
0.07 to 0.12M in THF
Samarium(II) iodide, Stabilized
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224-H260-H314-H318
Extremely flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c

H225-H302-H319-H335-H336-H351-H373

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ETHERS, N.O.S.

missing translation for 'einecsNumber' : 203-726-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only