missing translation for 'onlineSavingsMsg'
Learn More

Samarium cobalt, REacton™, Sm 33%

Product Code. 11335517
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335517 250 g 250g
11345517 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335517 Supplier Thermo Scientific Alfa Aesar Supplier No. 042732.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12017-68-4 | CoSm | 209.293 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12017-68-4
Molecular Formula CoSm
Molecular Weight (g/mol) 209.293
MDL Number MFCD02091682
InChI Key KPLQYGBQNPPQGA-UHFFFAOYSA-N
Synonym samarium cobalt, samarium cobalt powder,-30 mesh, cobalt-samarium, samarium-cobalt, powder30mesh, smco5, acmc-20ajrh, cobalt samarium, samarium cobalt, powder, sm2co17
PubChem CID 10125161
IUPAC Name cobalt;samarium
SMILES [Co].[Sm]

Specifications

Physical Form Fine Powder
Quantity 250 g
UN Number UN3089
Grade Reacton
Odor Odorless
Chemical Name or Material Samarium cobalt, Intermetallic, Magnet Alloy Additive

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.