missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Samarium, 99.9%, (trace metal basis), 20 to 40 mesh, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Boiling Point | 1778.0°C |
Melting Point | 1072.0°C |
Color | Gray |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.9% |
Linear Formula | Sm |
UN Number | 3132 |
Fieser | 17,305 |
Merck Index | 15, 8484 |
Show More |
Safety and Handling
Product Identifier
- Samarium, powder
- Danger
- Flammable solid Category 2
- Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
- H228-Flammable solid.
- H261-In contact with water releases flammable gases.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'