missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAM68 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102929
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KHDRBS1.
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| SAM68 | |
| Polyclonal | |
| Unconjugated | |
| KHDRBS1 | |
| FLJ34027; GAP-associated tyrosine phosphoprotein p62; GAP-associated tyrosine phosphoprotein p62 (Sam68); KH domain containing, RNA binding, signal transduction associated 1; KH domain-containing, RNA-binding, signal transduction-associated protein 1; KH RNA binding domain containing, signal transduction associated 1; KHDRBS1; khdrbs1 {ECO:0000312; p21 Ras GTPase-activating protein-associated p62; P62; p68; RGD:621459}; Sam68; src associated in mitosis, 68 kDa; src-associated in mitosis 68 kDa protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10657, 20218 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q07666, Q60749 | |
| KHDRBS1 | |
| A synthesized peptide derived from human KHDRBS1(Accession Q07666), corresponding to amino acid residues G392-R442. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction