missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAM68 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13434897
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13434897 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13434897 Supplier Invitrogen™ Supplier No. PA530400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: Jurakt, PC-12, Rat2. Predicted reactivity: Mouse (97%), Rat (97%), Pig (99%), Chicken (84%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAM68
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene KHDRBS1
Gene Accession No. Q07666, Q60749, Q91V33
Gene Alias FLJ34027; GAP-associated tyrosine phosphoprotein p62; GAP-associated tyrosine phosphoprotein p62 (Sam68); KH domain containing, RNA binding, signal transduction associated 1; KH domain-containing, RNA-binding, signal transduction-associated protein 1; KH RNA binding domain containing, signal transduction associated 1; KHDRBS1; khdrbs1 {ECO:0000312; p21 Ras GTPase-activating protein-associated p62; P62; p68; RGD:621459}; Sam68; src associated in mitosis, 68 kDa; src-associated in mitosis 68 kDa protein
Gene Symbols KHDRBS1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SAM68. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10657, 117268, 20218
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.