Learn More
Sam50 Mouse anti-Human, Unconjugated, Clone: 2H6H9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67425-1-IG-20UL
Description
The Sam50 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Sam50. This antibody recognizes human, pig, rat, mouse antigen. The Sam50 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Sam50 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 2H6H9 | |
| Unconjugated | |
| 20 μL | |
| 25813 | |
| IgG1 |