missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salvinorin B 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Salvinorin B |
| CAS | 92545-30-7 |
| Analyte or Component Names | Salvinorin B |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 O7 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Salvinorin B |
| InChI Formula | InChI=1S/C21H26O7/c1-20-6-4-12-19(25)28-15(11-5-7-27-10-11)9-21(12,2)17(20)16(23)14(22)8-13(20)18(24)26-3/h5,7,10,12-15,17,22H,4,6,8-9H2,1-3H3/t12-,13-,14-,15-,17-,20-,21-/m0/s1 |
| SMILES | COC(=O)[C@@H]1C[C@H](O)C(=O)[C@H]2[C@@]1(C)CC[C@H]3C(=O)O[C@@H](C[C@]23C)c4cocc4 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction