missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salvianolic acid B, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Description
Salvianolic acid B is an active ingredient of Salvia miltiorrhiza, which has been widely applied in China for the management of various microcirculation-related disorders, such as cardiovascular disease, cerebrovascular disease, and diabetic vascular complication.
Specifications
Specifications
| Chemical Name or Material | Salvianolic acid B |
| CAS | 121521-90-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C36H30O16 |
| Quantity | 5 mg |
| Synonym | Lithospermic acid B |
| Solubility Information | H2O : 50 mg/mL (69.58 mM; ultrasonic and adjust pH to 3 with HCl) ∣DMSO : 25 mg/mL (34.79 mM; Need ultrasonic) |
| SMILES | O=C([C@@H]1[C@@H](C2=CC=C(O)C(O)=C2)OC3=C(O)C=CC(/C=C/C(O[C@@H](C(O)=O)CC4=CC=C(O)C(O)=C4)=O)=C13)O[C@@H](C(O)=O)CC5=CC=C(O)C(O)=C5 |
| Molecular Weight (g/mol) | 718.61 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction