missing translation for 'onlineSavingsMsg'
Learn More

Salubrinal, TRC

Product Code. 30418005
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418005 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418005 Supplier Toronto Research Chemicals Supplier No. TRCS10055010MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Salubrinal
CAS 405060-95-9
Molecular Formula C21 H17 Cl3 N4 O S
Quantity 10 mg
Synonym (2E)-3-phenyl-N-[2,2,2-trichloro-1-[[(8-quinolinylamino)thioxomethyl]amino]ethyl]-2-propenamide
InChI Formula InChI=1S/C21H17Cl3N4OS/c22-21(23,24)19(27-17(29)12-11-14-6-2-1-3-7-14)28-20(30)26-16-10-4-8-15-9-5-13-25-18(15)16/h1-13,19H,(H,27,29)(H2,26,28,30)/b12-11+
SMILES ClC(Cl)(Cl)C(NC(=S)Nc1cccc2cccnc12)NC(=O)\C=C\c3ccccc3
IUPAC Name (E)-3-phenyl-N-[2,2,2-trichloro-1-(quinolin-8-ylcarbamothioylamino)ethyl]prop-2-enamide
Molecular Weight (g/mol) 479.81
Formula Weight 478.0189
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.