missing translation for 'onlineSavingsMsg'
Learn More

Salubrinal, MedChemExpress

Product Code. 16496776
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496776 100 mg 100mg
16493476 25 mg 25mg
16482657 50 mg 50mg
16466568 10 mg 10mg
16419147 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496776 Supplier Medchem Express Supplier No. HY15486
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Salubrinal is a cell-permeable and selective inhibitor of eIF2α dephosphorylation. Salubrinal acts as a dual-specificity phosphatase 2 (Dusp2) inhibitor and suppresses inflammation in anti-collagen antibody-induced arthritis. Salubrinal has antiviral activity against HSV-1 and inhibits dephosphorylation of eIF2α mediated by the HSV-1 protein ICP34.5.

Salubrinal is a cell-permeable and selective inhibitor of eIF2α dephosphorylation. Salubrinal acts as a dual-specificity phosphatase 2 (Dusp2) inhibitor and suppresses inflammation in anti-collagen antibody-induced arthritis. Salubrinal has antiviral activity against HSV-1 and inhibits dephosphorylation of eIF2α mediated by the HSV-1 protein ICP34.5.

Specifications

Chemical Name or Material Salubrinal
Physical Form Powder
CAS 405060-95-9
Color Light Brown
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H17Cl3N4OS
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 50 mg/mL (104.21 mM; Need ultrasonic)
SMILES O=C(NC(NC(NC1=CC=CC2=C1N=CC=C2)=S)C(Cl)(Cl)Cl)/C=C/C3=CC=CC=C3
Molecular Weight (g/mol) 479.81
Formula Weight 479.81
Percent Purity 99.5%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.