missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salsolinol-1-carboxylic acid, Tocris Bioscience™
Endogenous amino acid
Chemical Identifiers
CAS | 57256-34-5 |
---|---|
Molecular Formula | C11H13NO4 |
Molecular Weight (g/mol) | 223.228 |
InChI Key | XHGLVMDBZZZXDP-UHFFFAOYSA-N |
Synonym | 1-carboxysalsolinol, salsolinol-1-carboxylic acid, sal-1-ca, 1-methyl-6,7-dihydroxy-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid, 6,7-dihydroxy-1-methyl-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid, salsolinol-1-carboxylic acid_x000d_, 1-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-6,7-dihydroxy-1-methyl, 1-isoquinolinecarboxylicacid, 1,2,3,4-tetrahydro-6,7-dihydroxy-1-methyl, 6,7-dihydroxy-1-methyl-1,2,3,4-tetrahydro-1-isoquinolinecarboxylic acid #, isoquinoline-1-carboxylic acid, 1,2,3,4-tetrahydro-6,7-hydroxy-1-methyl |
PubChem CID | 100185 |
IUPAC Name | 6,7-dihydroxy-1-methyl-3,4-dihydro-2H-isoquinoline-1-carboxylic acid |
SMILES | CC1(C2=CC(=C(C=C2CCN1)O)O)C(=O)O |
Description
An amino acid that occurs naturally in the CNS.Chemical Identifiers
57256-34-5 | |
223.228 | |
1-carboxysalsolinol, salsolinol-1-carboxylic acid, sal-1-ca, 1-methyl-6,7-dihydroxy-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid, 6,7-dihydroxy-1-methyl-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid, salsolinol-1-carboxylic acid_x000d_, 1-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-6,7-dihydroxy-1-methyl, 1-isoquinolinecarboxylicacid, 1,2,3,4-tetrahydro-6,7-dihydroxy-1-methyl, 6,7-dihydroxy-1-methyl-1,2,3,4-tetrahydro-1-isoquinolinecarboxylic acid #, isoquinoline-1-carboxylic acid, 1,2,3,4-tetrahydro-6,7-hydroxy-1-methyl | |
6,7-dihydroxy-1-methyl-3,4-dihydro-2H-isoquinoline-1-carboxylic acid |
C11H13NO4 | |
XHGLVMDBZZZXDP-UHFFFAOYSA-N | |
100185 | |
CC1(C2=CC(=C(C=C2CCN1)O)O)C(=O)O |
Specifications
57256-34-5 | |
1-carboxysalsolinol, salsolinol-1-carboxylic acid, sal-1-ca, 1-methyl-6,7-dihydroxy-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid, 6,7-dihydroxy-1-methyl-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid, salsolinol-1-carboxylic acid_x000d_, 1-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-6,7-dihydroxy-1-methyl, 1-isoquinolinecarboxylicacid, 1,2,3,4-tetrahydro-6,7-dihydroxy-1-methyl, 6,7-dihydroxy-1-methyl-1,2,3,4-tetrahydro-1-isoquinolinecarboxylic acid #, isoquinoline-1-carboxylic acid, 1,2,3,4-tetrahydro-6,7-hydroxy-1-methyl | |
CC1(C2=CC(=C(C=C2CCN1)O)O)C(=O)O | |
223.228 | |
223.23 | |
Salsolinol-1-carboxylic acid |
C11H13NO4 | |
XHGLVMDBZZZXDP-UHFFFAOYSA-N | |
6,7-dihydroxy-1-methyl-3,4-dihydro-2H-isoquinoline-1-carboxylic acid | |
100185 | |
>98% |
Safety and Handling
Recommended Storage : Store at Room Temperature