missing translation for 'onlineSavingsMsg'
Learn More

Salsolidine, MedChemExpress

Product Code. 16477796
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477796 100 mg 100mg
16473736 25 mg 25mg
16463947 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16477796 Supplier Medchem Express Supplier No. HY22385
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Salsolidine is a tetrahydroisoquinoline alkaloid, acts as a stereoselective competitive MAO A inhibitor.

Salsolidine is a tetrahydroisoquinoline alkaloid, acts as a stereoselective competitive MAO A inhibitor.

Specifications

Chemical Name or Material Salsolidine
CAS 5784-74-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C12H17NO2
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (241.23 mM; Need ultrasonic)
SMILES CC1NCCC2=C1C=C(OC)C(OC)=C2
Molecular Weight (g/mol) 207.27
Formula Weight 207.27
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.