Learn More
Invitrogen™ Salmonella Polyclonal Antibody, HRP

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173021
Description
The PA1-73021 antibody reacts with Salmonella species. PA1-73021 has been successfully used in ELISA applications. The PA1-73021 antibody was raised against a mixture of S. enteriditis, S. typhimurium & S. heidelburg (O & H antigens). Recommended short term storage, up to 6 months, store at 4°C. For longer term storage, aliquot and freeze.
The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major surface antigens; the H or flagellar antigen (phase 1 and 2), the O or somatic antigen (part of the LPS moiety) and the Vi or capsular antigen (referred to as K in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram negative bacteria. This LPS is composed of an O polysaccharide (O antigen) an R core and the endotoxic inner Lipid A. Endotoxins evoke fever and can activate complement, kinin and clotting factors.
Specifications
| Salmonella | |
| Polyclonal | |
| HRP | |
| Salmonellae | |
| Mixture of S. enteriditis, S. typhimurium & S. heidelburg (O & H antigens). | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 10mg/mL BSA and 0.002% thimerosal; pH 7.2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.