Learn More
Thermo Scientific™ Salmonella enterica serovar poona NCTC 4840 Culti-Loop
Product Code. 12969068
Change view
Click to view available options
No. per Pack:
5 Pk.
Unit Size:
Pack of 5
Product Code. No. per Pack Unit Size
12969068 5 Pk. Pack of 5
1 options
If you would like to order, a signed ATCC agreement is required.
Product Code. 12969068 Supplier Thermo Scientific™ Supplier No. R4606009
  • ONLINE EXCLUSIVE PRICE
    £300.00
    £230.82 / Pack of 5
    Save £69.18
    23% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
If you would like to order, a signed ATCC agreement is required.
Culti-Loops containing the quality control microorganism Salmonella enterica serovar Poona NCTC 4840.
Thermo Scientific Oxoid Culti-Loops containing the quality control microorganism Salmonella enterica serovar poona NCTC 4840.
This product is a ready-to-use, disposable loop system designed for quality control in microbiology laboratories.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 4840
Certifications/Compliance 21 CFR Part 820
Manufacturing Origin United States
Product Type Culti-Loop
No. per Pack 5 Pk.
Species Salmonella enterica subsp. enterica serovar poona