missing translation for 'onlineSavingsMsg'
Learn More

Salmeterol Xinafoate, TRC

Product Code. 30009101
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009101 25 mg 25mg
30010493 50 mg 50mg
17982155 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009101 Supplier Toronto Research Chemicals Supplier No. TRCS09010025MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94749-08-3
Molecular Formula C25 H37 N O4 . C11 H8 O3
Molecular Weight (g/mol) 603.75
Synonym Salmeterol Xinafoate, (1RS)-1-[4-Hydroxy-3-(Hydroxymethyl)phenyl]-2-[[6-(4-phenylbutoxy)hexyl]amino]ethanol 1-hydroxynaphthalene-2-carboxylate, 2-Naphthalenecarboxylic acid, 1-hydroxy-, compd. with 4-hydroxy-α1-[[[6-(4-phenylbutoxy)hexyl]amino]methyl]-1,3-benzenedimethanol (1:1) (9CI, ACI), 1,3-Benzenedimethanol, 4-hydroxy-α1-[[[6-(4-phenylbutoxy)hexyl]amino]methyl]-, (±)-, 1-hydroxy-2-naphthalenecarboxylate (salt) (ZCI), 1,3-Benzenedimethanol, 4-hydroxy-α1-[[[6-(4-phenylbutoxy)hexyl]amino]methyl]-, 1-hydroxy-2-naphthalenecarboxylate (salt) (9CI), 2-Naphthalenecarboxylic acid, 1-hydroxy-, compd. with (±)-4-hydroxy-α1-[[[6-(4-phenylbutoxy)hexyl]amino]methyl]-1,3-benzenedimethanol (1:1) (ZCI), Arial, GR 33343G, GR 3343G, Salmetedur, Salmeterol hydroxynaphthoate, Serevent, Serevent Diskus, Siduomi, SN 408
IUPAC Name 2-(hydroxymethyl)-4-[1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]phenol;1-hydroxynaphthalene-2-carboxylic acid
SMILES OCc1cc(ccc1O)C(O)CNCCCCCCOCCCCc2ccccc2.OC(=O)c3ccc4ccccc4c3O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C25H37NO4.C11H8O3/c27-20-23-18-22(13-14-24(23)28)25(29)19-26-15-7-1-2-8-16-30-17-9-6-12-21-10-4-3-5-11-21;12-10-8-4-2-1-3-7(8)5-6-9(10)11(13)14/h3-5,10-11,13-14,18,25-29H,1-2,6-9,12,15-17,19-20H2;1-6,12H,(H,13,14)
Formula Weight 603.3196
Chemical Name or Material Salmeterol xinafoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.