missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Salmeterol xinafoate
GREENER_CHOICE

Product Code. 30001616
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001616 10 mg 10mg
30001628 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001616 Supplier Tocris Bioscience™ Supplier No. 4712/10
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Long-acting β2 agonist; bronchodilator

Long-acting β2 adrenergic receptor agonist. Inhibits the release of histamine, leukotrienes and prostaglandin D2, and blocks GM-CSF production from human cultured mast cells. Exhibits efficacious bronchodilatory activity in patients with asthma or chronic obstructive pulmonary disease (COPD).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94749-08-3
Molecular Formula C36H45NO7

Specifications

Quantity 10 mg
Formula Weight 603.75
Purity >99%
Molecular Weight (g/mol) 603.76
Chemical Name or Material Salmeterol xinafoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.